accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3NH3CNH3CCH3CH3H3CCH3(E/Z)(E/Z)

CAS RN 74663-77-7

IUPAC Name: N2,N3-bis[2,6-bis(propan-2-yl)phenyl]butane-2,3-diimine
Synonyms: N2,N3-bis(2,6-diisopropylphenyl)butane-2,3-diimine
Molecular Weight (g/mol): 404.64
Molecular Formula: C28H40N2
InChi Key: YUFQUBWPYIPRHZ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N=C(C)C(C)=NC1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results