accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNH

CAS RN 7467-91-6

IUPAC Name: 4,6-dihydroquinoxalin-6-one
Synonyms: 4H-quinoxalin-6-one
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: VEGOESHHZXUHMI-UHFFFAOYSA-N
SMILES: O=C1C=CC2=NC=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)