accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONO

CAS RN 7470-38-4

IUPAC Name: 4-(morpholin-4-yl)benzoic acid
Synonyms: 4-(morpholin-4-yl)benzoic acid
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: XVAJKPNTGSKZSQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results