accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7471-03-6

IUPAC Name: 2-amino-1,3-benzothiazol-4-ol
Synonyms: 4-benzothiazolol, 2-amino-
Molecular Weight (g/mol): 166.2
Molecular Formula: C7H6N2OS
InChi Key: PFQJPSASUCHKRO-UHFFFAOYSA-N
SMILES: NC1=NC2=C(O)C=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results