accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNCH3

CAS RN 747413-21-4

IUPAC Name: 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
Synonyms: 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine
Molecular Weight (g/mol): 302.23
Molecular Formula: C17H27BN2O2
InChi Key: RDFJBDMCBVSCFI-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results