accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSOOO

CAS RN 74801-29-9

IUPAC Name: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-(phenylsulfanyl)oxane
Synonyms: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-(phenylsulfanyl)oxane
Molecular Weight (g/mol): 632.82
Molecular Formula: C40H40O5S
InChi Key: IKCMSYGNAFDJNX-UHFFFAOYNA-N
SMILES: C(OCC1=CC=CC=C1)C1OC(SC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)