CAS RN 74807-44-6
IUPAC Name:
2-(2-aminoacetamido)-3-hydroxybutanoic acid dihydrate
Synonyms:
2-(2-aminoacetamido)-3-hydroxybutanoic acid dihydrate
Molecular Weight (g/mol):
212.2
Molecular Formula:
C6H16N2O6
InChi Key:
BFDXCPYYJSKNOU-UHFFFAOYNA-N
SMILES:
O.O.CC(O)C(NC(=O)CN)C(O)=O