accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7481-89-2

IUPAC Name: 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: zalcitabine|DDC
Molecular Weight (g/mol): 211.22
Molecular Formula: C9H13N3O3
InChi Key: WREGKURFCTUGRC-KGQMAECUNA-N
SMILES: NC1=NC(=O)N(C=C1)[C@H]1CC[C@@H](CO)O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results