accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNCl

CAS RN 748187-76-0

IUPAC Name: [(2-chloropyridin-4-yl)methyl](methyl)amine
Synonyms: [(2-chloropyridin-4-yl)methyl](methyl)amine
Molecular Weight (g/mol): 156.61
Molecular Formula: C7H9ClN2
InChi Key: NIBLXLUVASMQFM-UHFFFAOYSA-N
SMILES: CNCC1=CC(Cl)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results