accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74844-91-0

IUPAC Name: 1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 245.28
Molecular Formula: C11H19NO5
InChi Key: MZMNEDXVUJLQAF-SFYZADRCSA-N
SMILES: COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)