accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNH2

CAS RN 74852-62-3

IUPAC Name: 4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline
Synonyms: 4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline
Molecular Weight (g/mol): 283.38
Molecular Formula: C17H21N3O
InChi Key: VXEGSRKPIUDPQT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results