accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74866-28-7

IUPAC Name: 2,2'-dibromo-1,1'-binaphthalene
Synonyms: 2,2'-dibromo-1,1'-binaphthalene
Molecular Weight (g/mol): 412.12
Molecular Formula: C20H12Br2
InChi Key: IJUDEFZBMMRSNM-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC=CC2=C1C1=C(Br)C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)