CAS RN 7487-15-2
IUPAC Name:
1,4-dimethyl naphthalene-1,4-dicarboxylate
Synonyms:
1,4-dimethyl naphthalene-1,4-dicarboxylate
Molecular Weight (g/mol):
244.25
Molecular Formula:
C14H12O4
InChi Key:
OCSXMIBZIHMVCP-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C2C=CC=CC2=C(C=C1)C(=O)OC
1
–
1
of
1
results
1
Dimethyl 1,4-Naphthalenedicarboxylate 98.0+%, TCI America™
CAS: 7487-15-2 Molecular Formula: C14H12O4 Molecular Weight (g/mol): 244.246 MDL Number: MFCD04038075 InChI Key: OCSXMIBZIHMVCP-UHFFFAOYSA-N Synonym: 1,4-Naphthalenedicarboxylic Acid Dimethyl Ester PubChem CID: 11064621 IUPAC Name: dimethyl naphthalene-1,4-dicarboxylate SMILES: COC(=O)C1=CC=C(C2=CC=CC=C21)C(=O)OC