accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OOSOOZn

CAS RN 7488-52-0

IUPAC Name: zinc(2+) dihydrate sulfite
Synonyms: zinc(2+) dihydrate sulfite
Molecular Weight (g/mol): 181.47
Molecular Formula: H4O5SZn
InChi Key: QUPDPMXNPJYOTJ-UHFFFAOYSA-L
SMILES: O.O.[Zn++].[O-]S([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results