CAS RN 749151-25-5
IUPAC Name:
4-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]aniline
Synonyms:
4-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]aniline
Molecular Weight (g/mol):
282.36
Molecular Formula:
C16H14N2OS
InChi Key:
FEJBCMDICNNALW-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CSC(=N1)C1=CC=C(N)C=C1
1
–
1
of
1
results
1
2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole, 97%, Thermo Scientific™
CAS: 749151-25-5 Molecular Formula: C16H14N2OS Molecular Weight (g/mol): 282.361 InChI Key: FEJBCMDICNNALW-UHFFFAOYSA-N Synonym: 2-4-aminophenyl-4-4-methoxyphenyl thiazole,4-4-4-methoxyphenyl-1,3-thiazol-2-yl aniline,4-4-4-methoxyphenyl thiazol-2-yl aniline PubChem CID: 24261904 IUPAC Name: 4-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]aniline SMILES: COC1=CC=C(C=C1)C2=CSC(=N2)C3=CC=C(C=C3)N