CAS RN 7492-38-8
IUPAC Name:
2-methyloctan-4-one
Synonyms:
2-methyloctan-4-one
Molecular Weight (g/mol):
142.24
Molecular Formula:
C9H18O
InChi Key:
ICSKJDZASFIJQK-UHFFFAOYSA-N
SMILES:
CCCCC(=O)CC(C)C
1
–
1
of
1
results
1
Promotions Available
2-Methyl-4-octanone, 99%
2-Methyl-4-octanone, 99%
CAS: 7492-38-8 Molecular Formula: C9H18O Molecular Weight (g/mol): 142.24 MDL Number: MFCD00048808 InChI Key: ICSKJDZASFIJQK-UHFFFAOYSA-N Synonym: 2-methyl-4-octanone,4-octanone, 2-methyl PubChem CID: 522566 IUPAC Name: 2-methyloctan-4-one SMILES: CCCCC(=O)CC(C)C