accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74920-80-2

IUPAC Name: 4-{2-[4-(diethylamino)phenyl]diazen-1-yl}-1-methylpyridin-1-ium iodide
Synonyms: 4-{2-[4-(diethylamino)phenyl]diazen-1-yl}-1-methylpyridin-1-ium iodide
Molecular Weight (g/mol): 396.28
Molecular Formula: C16H21IN4
InChi Key: FBXMWVQMHYGKQY-UHFFFAOYSA-M
SMILES: [I-].CCN(CC)C1=CC=C(C=C1)N=NC1=CC=[N+](C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results