accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 7498-57-9

IUPAC Name: 2-(naphthalen-2-yl)acetonitrile
Synonyms: naphthalen-2-yl-acetonitrile
Molecular Weight (g/mol): 167.21
Molecular Formula: C12H9N
InChi Key: LPCWDVLDJVZIHA-UHFFFAOYSA-N
SMILES: N#CCC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)