accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7499-08-3

IUPAC Name: 3-chloro-2-methylbenzoic acid
Synonyms: 3-chloro-2-methylbenzoic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: HXGHMCLCSPQMOR-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results