accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 749927-69-3

IUPAC Name: 4-bromo-2-fluoro-N-methylbenzamide
Synonyms: 4-bromo-2-fluoro-N-methylbenzamide
Molecular Weight (g/mol): 232.05
Molecular Formula: C8H7BrFNO
InChi Key: BAJCFNRLEJHPTQ-UHFFFAOYSA-N
SMILES: CNC(=O)C1=C(F)C=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results