accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3PH3CCH3H3CPCH3PH3CCH3H3CNHCH3HClClClHRuClHHClRu(?)(?)

CAS RN 749935-02-2

IUPAC Name: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane) hydrochloride ruthenium chloride
Synonyms: [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane) hydrochloride ruthenium chloride
Molecular Weight (g/mol): 1786.1
Molecular Formula: C98H91Cl5NP4Ru2
InChi Key: BKLYSQLZSZMPAP-UHFFFAOYSA-M
SMILES: Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)