CAS RN 75-01-4
CAS RN 75-01-4
IUPAC Name:
chloroethene
Synonyms:
vinyl chloride vinylchlorid vinile (cloruro di) cloroetileno chlorure de vinyle trovidur vinyl chloride monomer VCM vinyl chlorine ccris 621
Molecular Weight (g/mol):
62.5
Molecular Formula:
C2H3Cl
InChi Key:
BZHJMEDXRYGGRV-UHFFFAOYSA-N
SMILES:
ClC=C
1
–
1
of
1
results
1
Vinyl Chloride, SPEX CertiPrep™
CAS: 75-01-4 Molecular Formula: C2H3Cl Molecular Weight (g/mol): 62.496 InChI Key: BZHJMEDXRYGGRV-UHFFFAOYSA-N PubChem CID: 6338 ChEBI: CHEBI:28509 IUPAC Name: chloroethene SMILES: C=CCl