accessibility menu, dialog, popup

UserName

Viewing profile as user:
NCH3H3CCH3BH3

CAS RN 75-22-9

IUPAC Name: trimethylamine borane
Synonyms: trimethylamine borane
Molecular Weight (g/mol): 72.95
Molecular Formula: C3H12BN
InChi Key: WVMHLYQJPRXKLC-UHFFFAOYSA-N
SMILES: B.CN(C)C
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (4)
  • (1)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)