accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2FBFF

CAS RN 75-23-0

IUPAC Name: ethanamine; trifluoroborane
Synonyms: boron trifluoride; ethylamine
Molecular Weight (g/mol): 112.89
Molecular Formula: C2H7BF3N
InChi Key: JDVIRCVIXCMTPU-UHFFFAOYSA-N
SMILES: CCN.FB(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results