accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-28-5

IUPAC Name: 2-methylpropane
Synonyms: isobutane caswell no. 503A 0,1% w/w butadiene A 31 (hydrocarbon) epa pesticide chemical code 097101
Molecular Weight (g/mol): 58.12
Molecular Formula: C4H10
InChi Key: NNPPMTNAJDCUHE-UHFFFAOYSA-N
SMILES: CC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
Technique 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results