accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-31-0

IUPAC Name: propan-2-amine
Synonyms: isopropylamine isopropilamina 2-amino-propano
Molecular Weight (g/mol): 59.11
Molecular Formula: C3H9N
InChi Key: JJWLVOIRVHMVIS-UHFFFAOYSA-N
SMILES: CC(C)N
Boiling Point 
  • (1)
  • (5)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (9)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (5)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (9)
  • (3)
  • (1)
  • (2)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)