accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClCl

CAS RN 75-34-3

IUPAC Name: 1,1-dichloroethane
Synonyms: 1,1-dichloroethane
Molecular Weight (g/mol): 98.95
Molecular Formula: C2H4Cl2
InChi Key: SCYULBFZEHDVBN-UHFFFAOYSA-N
SMILES: CC(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results