accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 75-56-9

IUPAC Name: 2-methyloxirane
Synonyms: propylene oxide epoxy-propane propylene epoxide epoxypropane caswell no. 713A 1,2-propylene oxide epa pesticide chemical code 042501 AD 6 (suspending agent) ccris 540 propane, epoxy-
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: GOOHAUXETOMSMM-UHFFFAOYNA-N
SMILES: CC1CO1
Percent Purity 
  • (7)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
Boiling Point 
  • (8)
  • (3)
  • (7)
Quantity 
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (12)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (6)
  • (1)
  • (2)
  • (10)
  • (7)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (8)
  • (3)
  • (7)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (12)
  • (2)
  • (5)
  • (2)
  • (1)