accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75-58-1

IUPAC Name: tetramethylazanium iodide
Synonyms: tetramethylammonium iodide
Molecular Weight (g/mol): 201.05
Molecular Formula: C4H12IN
InChi Key: RXMRGBVLCSYIBO-UHFFFAOYSA-M
SMILES: [I-].C[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)