accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H2N

CAS RN 75-64-9

IUPAC Name: 2-methylpropan-2-amine
Synonyms: erbumine trimethylaminomethane T-butylamine butylamine, tert tert butylamine tertiary butylamine butylamine, tertiary tertiary-butylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: YBRBMKDOPFTVDT-UHFFFAOYSA-N
SMILES: CC(C)(C)N
Molecular Weight (g/mol) 
  • (6)
  • (7)
  • (7)
Boiling Point 
  • (4)
  • (3)
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (8)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (6)
  • (1)
  • (3)
  • (10)
  • (6)
  • (7)
  • (7)
  • (4)
  • (3)
  • (6)
  • (6)
  • (3)
  • (2)
  • (8)
  • (6)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (7)
  • (1)