accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClClCl

CAS RN 75-69-4

IUPAC Name: trichloro(fluoro)methane
Synonyms: trichlorofluoromethane eskimon 11 freon 11
Molecular Weight (g/mol): 137.36
Molecular Formula: CCl3F
InChi Key: CYRMSUTZVYGINF-UHFFFAOYSA-N
SMILES: FC(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
Concentration 
  • (1)
Element 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)