CAS RN 75-83-2
IUPAC Name:
2,2-dimethylbutane
Synonyms:
2,2-dimethylbutane
Molecular Weight (g/mol):
86.18
Molecular Formula:
C6H14
InChi Key:
HNRMPXKDFBEGFZ-UHFFFAOYSA-N
SMILES:
CCC(C)(C)C
1
–
1
of
1
results
1
2,2-Dimethylbutane, SPEX CertiPrep™
CAS: 75-83-2 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 InChI Key: HNRMPXKDFBEGFZ-UHFFFAOYSA-N PubChem CID: 6403 IUPAC Name: 2,2-dimethylbutane SMILES: CCC(C)(C)C