accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOO

CAS RN 7500-53-0

IUPAC Name: 2-[2-(carboxymethyl)phenyl]acetic acid
Synonyms: 1,2-benzenediacetic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: MMEDJBFVJUFIDD-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1CC(O)=O
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)