accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7501-37-3

IUPAC Name: 4-[4-(propan-2-yl)phenyl]butanoic acid
Synonyms: 4-(4-isopropylphenyl)butanoic acid
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: COMUZXZDTNYSJE-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(CCCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results