accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHNN

CAS RN 7504-94-1

IUPAC Name: 2-hydrazinylpyrimidine
Synonyms: 2-hydrazinylpyrimidine
Molecular Weight (g/mol): 110.12
Molecular Formula: C4H6N4
InChi Key: QDGHXQFTWKRQTG-UHFFFAOYSA-N
SMILES: NNC1=NC=CC=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results