accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 750573-24-1

IUPAC Name: 9-[3-bromo-5-(9H-carbazol-9-yl)phenyl]-9H-carbazole
Synonyms: 9-[3-bromo-5-(carbazol-9-yl)phenyl]carbazole
Molecular Weight (g/mol): 487.4
Molecular Formula: C30H19BrN2
InChi Key: SJOKONNBSXFPSN-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results