accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrBr

CAS RN 750573-26-3

IUPAC Name: 9-(3,5-dibromophenyl)-9H-carbazole
Synonyms: 9-(3,5-dibromophenyl)carbazole
Molecular Weight (g/mol): 401.1
Molecular Formula: C18H11Br2N
InChi Key: YXHXRBIAIVNCPG-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(Br)=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results