accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNIOOH

CAS RN 75092-30-7

IUPAC Name: 4-iodo-1-methyl-1H-pyrazole-5-carboxylic acid
Synonyms: 4-iodo-2-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 252.01
Molecular Formula: C5H5IN2O2
InChi Key: SZRMOKQNOJYMNK-UHFFFAOYSA-N
SMILES: CN1N=CC(I)=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results