accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 751-38-2

IUPAC Name: 1,2,3,4-tetraphenylnaphthalene
Synonyms: 1,2,3,4-tetraphenylnaphthalene
Molecular Weight (g/mol): 432.57
Molecular Formula: C34H24
InChi Key: UCTTYTFENYGAPP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C(C2=CC=CC=C2)C(C2=CC=CC=C2)=C(C2=CC=CC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results