CAS RN 75187-63-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PFKFB3 Inhibitor III, YN1
A cell-permeable, potent and competitive inhibitor of fructose-2,6-bisphosphatase 3 (PFKFB3; IC50 = 670 nM).