accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNOBr

CAS RN 7520-95-8

IUPAC Name: 2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethan-1-one
Synonyms: 2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethanone
Molecular Weight (g/mol): 296.18
Molecular Formula: C12H10BrNOS
InChi Key: BOMSILGSXFNLJX-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=CC=C1)C(=O)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results