accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7521-41-7

IUPAC Name: 2-amino-3-formylpyridin-1-ium
Synonyms: 2-amino-3-formylpyridin-1-ium
Molecular Weight (g/mol): 123.13
Molecular Formula: C6H7N2O
InChi Key: NXMFJCRMSDRXLD-UHFFFAOYSA-O
SMILES: NC1=[NH+]C=CC=C1C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)