accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 753-90-2

IUPAC Name: 2,2,2-trifluoroethan-1-amine
Synonyms: 2,2,2-trifluoroethylamine
Molecular Weight (g/mol): 99.06
Molecular Formula: C2H4F3N
InChi Key: KIPSRYDSZQRPEA-UHFFFAOYSA-N
SMILES: NCC(F)(F)F
Boiling Point 
  • (4)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (6)
  • (4)
  • (3)
  • (2)
  • (3)
  • (5)
  • (7)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)