accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NH2OH

CAS RN 7533-40-6

IUPAC Name: (2S)-2-amino-4-methylpentan-1-ol
Synonyms: L-leucinol
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: VPSSPAXIFBTOHY-LURJTMIESA-N
SMILES: CC(C)C[C@H](N)CO
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)