accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75336-86-6

IUPAC Name: (2R)-2-methylpiperazine
Synonyms: (2R)-2-methylpiperazine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: JOMNTHCQHJPVAZ-RXMQYKEDSA-N
SMILES: C[C@@H]1CNCCN1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)