accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7534-94-3

IUPAC Name: 1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl 2-methylprop-2-enoate
Synonyms: isobornyl methacrylate
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: IAXXETNIOYFMLW-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)