accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNOOOHO

CAS RN 7536-58-5

IUPAC Name: 4-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C16H21NO6
InChi Key: SOHLZANWVLCPHK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)