accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NH2O

CAS RN 754-10-9

IUPAC Name: 2,2-dimethylpropanamide
Synonyms: pivalamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: XIPFMBOWZXULIA-UHFFFAOYSA-N
SMILES: CC(C)(C)C(N)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (5)