accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 754214-56-7

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 244.1
Molecular Formula: C13H17BN2O2
InChi Key: UOXAMYZTYZLSCC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)