accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 754226-39-6

IUPAC Name: 2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 240.06
Molecular Formula: C12H15BF2O2
InChi Key: VJTCPZZOSKBSCP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results